CH4424847714 ChemBridge Corporation

4-amino-N,4-dimethylpentanamide hydrate

  • MDL number MFCD26959427
  • Cas Number : 1609406-99-6
  • Molecular Weight : 162.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data H2O
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4041154

Safety Information

MSDS https://www.hit2lead.com/sds/4041154_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart