CH4424847715 ChemBridge Corporation

3-(4-fluorophenoxy)azetidine hydrochloride

  • MDL number MFCD05664839
  • Cas Number : 63843-78-7
  • Molecular Weight : 203.6411

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4041166

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4041166
Availability details for
Quantity
  • Availability
  • Add to Cart