CH4424847716 ChemBridge Corporation

1-[2-(methylthio)ethyl]piperazine dihydrochloride

  • MDL number MFCD16620372
  • Cas Number : 88708-43-4
  • Molecular Weight : 233.2022

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4041175

Safety Information

MSDS https://www.hit2lead.com/sds/4041175_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart