CH4424847717 ChemBridge Corporation

2-propylproline hydrochloride

  • MDL number MFCD19103484
  • Cas Number : 2103397-17-5
  • Molecular Weight : 193.6711

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4041181

Safety Information

MSDS https://www.hit2lead.com/sds/4041181_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart