CH4424847724 ChemBridge Corporation

1-[(6-chloro-3-pyridinyl)methyl]piperazine dihydrochloride

  • MDL number MFCD09607616
  • Cas Number : 1185312-79-1
  • Molecular Weight : 284.6131

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4041330

Safety Information

MSDS https://www.hit2lead.com/sds/4041330_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart