CH4424847727 ChemBridge Corporation

N-methyl-2-piperidinecarboxamide hydrochloride

  • MDL number MFCD13562341
  • Cas Number : 1236263-43-6
  • Molecular Weight : 178.6598

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4041354

Safety Information

MSDS https://www.hit2lead.com/sds/4041354_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart