CH4424847748 ChemBridge Corporation

1-methyl-L-proline hydrochloride

  • MDL number MFCD06795802
  • Cas Number : 89614-97-1
  • Molecular Weight : 165.618

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4030671

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4030671
Availability details for
Quantity
  • Availability
  • Add to Cart