CH4424847756 ChemBridge Corporation

[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]amine dihydrochloride

  • MDL number MFCD12026975
  • Cas Number : 1158207-20-5
  • Molecular Weight : 239.1421

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4030682

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4030682
Availability details for
Quantity
  • Availability
  • Add to Cart