CH4424847761 ChemBridge Corporation

1-(4-morpholinylcarbonyl)-4-piperidinamine hydrochloride

  • MDL number MFCD15202379
  • Cas Number : 1211474-97-3
  • Molecular Weight : 249.7377

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4042569

Safety Information

MSDS https://www.hit2lead.com/sds/4042569_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart