CH4424847762 ChemBridge Corporation

N'-hydroxy-5-methyl-2-furancarboximidamide hydrochloride

  • MDL number MFCD22666485
  • Cas Number : 1390739-62-4
  • Molecular Weight : 176.6009

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4042580

Safety Information

MSDS https://www.hit2lead.com/sds/4042580_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart