CH4424847785 ChemBridge Corporation

methyl 3-bromoimidazo[1,2-a]pyridine-2-carboxylate dihydrobromide

  • MDL number MFCD26959402
  • Cas Number : 1609406-64-5
  • Molecular Weight : 416.892

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4042903

Safety Information

MSDS https://www.hit2lead.com/sds/4042903_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart