CH4424847786 ChemBridge Corporation

methyl 4-ethyl-4-piperidinecarboxylate hydrochloride

  • MDL number MFCD12408565
  • Cas Number : 1205749-08-1
  • Molecular Weight : 207.6977

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4042910

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4042910
Availability details for
Quantity
  • Availability
  • Add to Cart