CH4424847802 ChemBridge Corporation

(2,2-dimethylpropyl)hydrazine dihydrochloride

  • MDL number MFCD09834107
  • Cas Number : 1177323-30-6
  • Molecular Weight : 175.0999

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4043230

Safety Information

MSDS https://www.hit2lead.com/sds/4043230_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart