CH4424847804 ChemBridge Corporation

3-ethyl-3-methylpiperidine hydrochloride

  • MDL number MFCD26959469
  • Cas Number : 1609403-36-2
  • Molecular Weight : 163.6882

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4043267

Safety Information

MSDS https://www.hit2lead.com/sds/4043267_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart