CH4424847818 ChemBridge Corporation

3-(chloromethyl)-5-ethyl-1H-1,2,4-triazole hydrochloride

  • MDL number MFCD24842856
  • Cas Number : 1443980-82-2
  • Molecular Weight : 182.0511

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4043603

Safety Information

MSDS https://www.hit2lead.com/sds/4043603_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart