CH4424847826 ChemBridge Corporation

3-amino-7-ethyl-1,3-dihydro-2H-indol-2-one hydrochloride

  • MDL number MFCD19103304
  • Cas Number : 1332528-28-5
  • Molecular Weight : 212.676

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4043698

Safety Information

MSDS https://www.hit2lead.com/sds/4043698_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart