CH4424847836 ChemBridge Corporation

1-(1-piperidinylcarbonyl)-4-piperidinamine hydrochloride

  • MDL number MFCD18071273
  • Cas Number : 651056-87-0
  • Molecular Weight : 247.7649

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4043819

Safety Information

MSDS https://www.hit2lead.com/sds/4043819_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart