CH4424847861 ChemBridge Corporation

4-ethyl-3-methyl-2-piperazinone hydrochloride

  • MDL number MFCD22422047
  • Cas Number : 1417566-35-8
  • Molecular Weight : 178.6598

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4044255

Safety Information

MSDS https://www.hit2lead.com/sds/4044255_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart