CH4424847872 ChemBridge Corporation

4-ethyl-2-piperazinone hydrochloride

  • MDL number MFCD22422017
  • Cas Number : 65464-00-8
  • Molecular Weight : 164.6332

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4044495

Safety Information

MSDS https://www.hit2lead.com/sds/4044495_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart