CH4424847899 ChemBridge Corporation

1-(3-chloro-2-pyridinyl)piperazine dihydrochloride

  • MDL number MFCD12197241
  • Cas Number : 1193389-39-7
  • Molecular Weight : 270.5866

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4045142

Safety Information

MSDS https://www.hit2lead.com/sds/4045142_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart