CH4424847946 ChemBridge Corporation

4-(3-piperidinyl)pyrimidine dihydrochloride

  • MDL number MFCD12028541
  • Cas Number : 1185105-86-5
  • Molecular Weight : 236.1415

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4045489

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4045489
Availability details for
Quantity
  • Availability
  • Add to Cart