CH4424847975 ChemBridge Corporation

[(2-methylphenyl)(2-thienyl)methyl]amine hydrochloride

  • MDL number MFCD18483477
  • Cas Number : 1269376-69-3
  • Molecular Weight : 239.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://www.hit2lead.com/building-blocks/4031269

Safety Information

msds https://www.hit2lead.com/building-blocks/4031269
Availability details for
Quantity
  • Availability
  • Add to Cart