CH4424848115 ChemBridge Corporation

cis-2-aminocyclobutanol hydrochloride

  • MDL number MFCD26127763
  • Cas Number : 2070860-49-8
  • Molecular Weight : 123.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data HCl
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4068939

Safety Information

MSDS https://www.hit2lead.com/sds/4068939_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart