CH4424848214 ChemBridge Corporation

N-methyl-1-(1-methyl-2-piperidinyl)methanamine oxalate (1:2)

  • MDL number MFCD11841320
  • Cas Number : 1185121-07-6
  • Molecular Weight : 232.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data 2(COOH)2
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4088042

Safety Information

MSDS https://www.hit2lead.com/sds/4088042_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart