CH4424848218 ChemBridge Corporation

1-(3-methoxybenzyl)piperazine bis(trifluoroacetate)

  • MDL number MFCD11841335
  • Cas Number : 1185004-39-0
  • Molecular Weight : 434.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data 2CF3COOH
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://www.hit2lead.com/building-blocks/4088051

Safety Information

msds https://www.hit2lead.com/building-blocks/4088051
Availability details for
Quantity
  • Availability
  • Add to Cart