CH4424849269 ChemBridge Corporation

2H-1,2,3-triazol-4-amine

  • MDL number MFCD20502555
  • Cas Number : 573713-80-1
  • Molecular Weight : 84.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4089216

Safety Information

MSDS https://www.hit2lead.com/sds/4089216_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart