CH4424850319 ChemBridge Corporation

N-(2-phenylethyl)cyclohexanamine

  • MDL number MFCD00961593
  • Cas Number : 51827-40-8
  • Molecular Weight : 203.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4090394

Safety Information

MSDS https://www.hit2lead.com/sds/4090394_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart