CH4424850646 ChemBridge Corporation

3-(4-chloro-1H-pyrazol-1-yl)-1-propanamine

  • MDL number MFCD03419760
  • Cas Number : 1006447-90-0
  • Molecular Weight : 159.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Storage temp. -20
Physical Form liquid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4090779

Safety Information

MSDS https://www.hit2lead.com/sds/4090779_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart