CH4424851020 ChemBridge Corporation

ethyl 1,4'-bipiperidine-3-carboxylate dihydrochloride

  • MDL number MFCD08448123
  • Cas Number : 864292-95-5
  • Molecular Weight : 313.2637

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4100006

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4100006
Availability details for
Quantity
  • Availability
  • Add to Cart