CH4424851063 ChemBridge Corporation

1-(4-pyridinylmethyl)piperazine bis(trifluoroacetate)

  • MDL number MFCD28024823
  • Cas Number : 1609406-76-9
  • Molecular Weight : 405.2929

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2CF3COOH
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4101649

Safety Information

MSDS https://www.hit2lead.com/sds/4101649_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart