CH4424851065 ChemBridge Corporation

1H-pyrazol-3-ylboronic acid dihydrate

  • MDL number MFCD28024827
  • Molecular Weight : 147.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data 2H2O
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4201615

Safety Information

signalword Warning
MSDS https://www.hit2lead.com/sds/4201615_sds.pdf
hstatements H302, H315, H319, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart