CH4424851073 ChemBridge Corporation

N-(2-methoxybenzyl)-1-adamantanamine hydrobromide

  • MDL number MFCD13186496
  • Cas Number : 1609406-30-5
  • Molecular Weight : 352.3091

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5562419

Safety Information

MSDS https://www.hit2lead.com/sds/5562419_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart