CH4424851075 ChemBridge Corporation

[3-(2H-tetrazol-2-yl)-1-adamantyl]amine hydrochloride hydrate

  • MDL number MFCD26959743
  • Molecular Weight : 273.7624

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl H2O
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5627729

Safety Information

MSDS https://www.hit2lead.com/sds/5627729_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart