CH4424851076 ChemBridge Corporation

1-benzyl-4-(4-piperidinyl)piperazine trihydrochloride

  • MDL number MFCD28024833
  • Cas Number : 1609409-31-5
  • Molecular Weight : 368.7726

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 3HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6782149

Safety Information

MSDS https://www.hit2lead.com/sds/6782149_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart