CH4424851084 ChemBridge Corporation

[2,2,2-trifluoro-1-(3-pyridinyl)ethyl]amine dihydrochloride

  • MDL number MFCD06738892
  • Cas Number : 1159825-86-1
  • Molecular Weight : 249.061

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4100719

Safety Information

MSDS https://www.hit2lead.com/sds/4100719_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart