CH4424851094 ChemBridge Corporation

(3-thienylmethyl)amine hydrochloride

  • MDL number MFCD03306024
  • Cas Number : 115132-84-8
  • Molecular Weight : 149.6417

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4100798

Safety Information

MSDS https://www.hit2lead.com/sds/4100798_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart