CH4424851107 ChemBridge Corporation

rac-(2R,6R)-2,6-diallyl-1,2,3,6-tetrahydropyridine hydrochloride

  • MDL number MFCD00464920
  • Cas Number : 138617-50-2
  • Molecular Weight : 199.7203

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4100925

Safety Information

MSDS https://www.hit2lead.com/sds/4100925_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart