CH4424851113 ChemBridge Corporation

1-ethyl-N-[2-(2-pyridinyl)ethyl]-4-piperidinamine tri[(2Z)-2-butenedioate]

  • MDL number MFCD26959623
  • Cas Number : 1609430-40-1
  • Molecular Weight : 581.569

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 3cis-HOOCCH=CHCOOH
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4100991

Safety Information

MSDS https://www.hit2lead.com/sds/4100991_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart