CH4424851118 ChemBridge Corporation

(1,3-benzoxazol-2-ylmethyl)amine hydrochloride

  • MDL number MFCD09763577
  • Cas Number : 1072806-65-5
  • Molecular Weight : 184.6229

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4101049

Safety Information

MSDS https://www.hit2lead.com/sds/4101049_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart