CH4424851139 ChemBridge Corporation

(1H-benzimidazol-2-ylmethyl)amine dihydrochloride hydrate

  • MDL number MFCD00150073
  • Cas Number : 1155659-33-8
  • Molecular Weight : 238.1143

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl H2O
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4100108

Safety Information

MSDS https://www.hit2lead.com/sds/4100108_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart