CH4424851149 ChemBridge Corporation

[(1-methyl-1H-imidazol-2-yl)methyl]amine dihydrochloride

  • MDL number MFCD09431497
  • Cas Number : 53332-67-5
  • Molecular Weight : 184.0669

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4101359

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4101359
Availability details for
Quantity
  • Availability
  • Add to Cart