CH4424851159 ChemBridge Corporation

[2-(1H-pyrazol-4-yl)ethyl]amine dihydrochloride

  • MDL number MFCD09800617
  • Molecular Weight : 184.0669

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4101520

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4101520
Availability details for
Quantity
  • Availability
  • Add to Cart