CH4424851168 ChemBridge Corporation

1-(2-thienylmethyl)piperazine dihydrochloride

  • MDL number MFCD04116377
  • Cas Number : 6803-90-3
  • Molecular Weight : 255.2077

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4101650

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4101650
Availability details for
Quantity
  • Availability
  • Add to Cart