CH4424851202 ChemBridge Corporation

(4'-methyl-4-biphenylyl)amine hydrochloride

  • MDL number MFCD04971967
  • Cas Number : 811842-54-3
  • Molecular Weight : 219.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4102391

Safety Information

MSDS https://www.hit2lead.com/sds/4102391_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart