CH4424851203 ChemBridge Corporation

(2'-methyl-4-biphenylyl)amine hydrochloride

  • MDL number MFCD06739442
  • Cas Number : 811842-48-5
  • Molecular Weight : 219.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4102393

Safety Information

MSDS https://www.hit2lead.com/sds/4102393_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart