CH4424851206 ChemBridge Corporation

(3'-chloro-4-biphenylyl)amine hydrochloride

  • MDL number MFCD06739433
  • Cas Number : 811842-60-1
  • Molecular Weight : 240.1284

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4102403

Safety Information

MSDS https://www.hit2lead.com/sds/4102403_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart