CH4424851214 ChemBridge Corporation

[1-(1-adamantyl)-1-methylethyl]amine hydrochloride

  • MDL number MFCD02208075
  • Cas Number : 1660-17-9
  • Molecular Weight : 229.7894

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4102457

Safety Information

MSDS https://www.hit2lead.com/sds/4102457_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart