CH4424851229 ChemBridge Corporation

(6-methoxy-1,2,3,4-tetrahydro-1-naphthalenyl)amine hydrochloride

  • MDL number MFCD01712013
  • Cas Number : 39226-88-5
  • Molecular Weight : 213.7038

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4102674

Safety Information

MSDS https://www.hit2lead.com/sds/4102674_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart