CH4424851231 ChemBridge Corporation

[1-(2-chlorophenyl)ethyl]amine hydrochloride

  • MDL number MFCD11841333
  • Cas Number : 856629-37-3
  • Molecular Weight : 192.0856

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4102676

Safety Information

MSDS https://www.hit2lead.com/sds/4102676_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart