CH4424851236 ChemBridge Corporation

N-methyl-1-(4-pyrimidinyl)methanamine dihydrochloride

  • MDL number MFCD07781110
  • Cas Number : 1984118-01-5
  • Molecular Weight : 196.0776

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4102726

Safety Information

MSDS https://www.hit2lead.com/sds/4102726_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart